missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone-lysine N-methyltransferase SETDB1 Protein

A DNA sequence encoding the Homo sapiens (Human) Histone-lysine N-methyltransferase SETDB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7573-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16059490

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001138887.1
Histone-lysine N-methyltransferase SETDB1 Protein
Research Use Only
SETDB1
Recombinant Protein
E. coli
MSSLPGCIGL DAATATVESE EIAELQQAVV EELGISMEEL RHFIDEELEK MDCVQQRKKQ LAELETWVIQ KESEVAHVDQ LFDDASRAVT NCESLVKDFY SKLGLQYRDS SSEDESSRPT EIIEIPDEDD DVLSIDSGDA GSRTPKDQKL REAMAALRKS AQDVQKFMDA VNKKSSSQDL HKGTLSQMSG ELSKDGDLIV SMRILGKKRT KTWHKGTLIA IQTVGPGKKY KVKFDNKGKS LLSGNHIAYD YHPPADKLYV GSRVVAKYKD GNQVWLYAGI VAETPNVKNK LRFLIFFDDG YASYVTQSEL YPICRPLKKT WEDIEDISCR DFIEEYVTAY PNRPMVLLKS GQLIKTEWEG TWWKSRVEEV DGSLVRILFL VLFFSTILEA EVGGGGT
Greater than 80% as determined by SDS-PAGE.
9869
50 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers