missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HLA-DQA2 Protein

A DNA sequence encoding the Homo sapiens (Human) HLA-DQA2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8816-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16056641

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_064440.1
HLA-DQA2 Protein
200 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
3118
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HLA-DQA2
Recombinant Protein
E. coli
EDIVADHVAS YGVNFYQSHG PSGQYTHEFD GDEEFYVDLE TKETVWQLPM FSKFISFDPQ SALRNMAVGK HTLEFMMRQS NSTAATNEVP EVTVFSKFPV TLGQPNTLIC LVDNIFPPVV NITWLSNGHS VTEGVSETSF LSKSDHSFFK ISYLTFLPSA DEIYDCKVEH WGLDEPLLKH WEPEIPAPMS E
Videos
SDS
Documents