Läs mer
enQuireBio™ Recombinant Human HLA-F Protein
Produktkod. 15919918
Kvantitet:
10 μg
100 μg
2 μg
Förpackningsstorlek:
100µg
10µg
2µg
Produktkod. 15919918
Brand: enQuireBio™ QP122772ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
A cDNA sequence encoding the HLA-F was constructed and used to recombinantly synthesize the protein.
Specifikationer
Gen-ID (Entrez) 3134
Namn HLA-F Protein
Kvantitet 2 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gensymbol HLA-F
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His
Sekvens MGSSHHHHHH SSGLVPRGSH MGSGSHSLRY FSTAVSRPGR GEPRYIAVEY VDDTQFLRFD SDAAIPRMEP REPWVEQEGP QYWEWTTGYA KANAQTDRVA LRNLLRRYNQ SEAGSHTLQG MNGCDMGPDG RLLRGYHQHA YDGKDYISLN EDLRSWTAAD TVAQITQRFY EAEEYAEEFR TYLEGECLEL LRRYLENGKE TLQRADPPKA HVAHHPISDH EATLRCWALG FYPAEITLTW QRDGEEQTQD TELVETRPAG DGTFQKWAAV VVPPGEEQRY TCHVQHEGLP QPLILRWEQS PQPTIPI
Buffert HLA-F protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl 20% glycerol and 1mM DTT.
Renhet eller kvalitetsklass Greater than 90.0% as determined by SDS-PAGE.