missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HMGCR Protein
A DNA sequence encoding the Homo sapiens (Human) HMGCR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6169-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000850.1 | |
HMGCR Protein | |
Research Use Only | |
HMGCR | |
Recombinant Protein | |
E. coli | |
MTRGPVVRLP RACDSAEVKA WLETSEGFAV IKEAFDSTSR FARLQKLHTS IAGRNLYIRF QSRSGDAMGM NMISKGTEKA LSKLHEYFPE MQILAVSGNY CTDKKPAAIN WIEGRGKSVV CEAVIPAKVV REVLKTTTEA MIEVNINKNL VGSAMAGSIG GYNAHAANIV TAIYIACGQD AAQNVGSSNC ITLMEASGPT NEDLYISCTM PSIEIGTVGG GTNLLPQQAC LQMLGVQGAC KDNPGENARQ LARIVCGTVM AGELSLMAAL AAGHLVKSHM IHNRSKINLQ DLQGACTKKT | |
Greater than 90% as determined by SDS-PAGE. |
3156 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |