Läs mer
enQuireBio™ Recombinant Human HMOX2 Protein
Produktkod. p-7150145
Kvantitet:
1 mg
25 μg
5 μg
Förpackningsstorlek:
1mg
25µg
5µg
Produktkod. 15919948
Brand: enQuireBio™ QP122875ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
A cDNA sequence encoding the HMOX2 was constructed and used to recombinantly synthesize the protein.
Specifikationer
Namn HMOX2 Protein
Kvantitet 5 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag Untagged
Sekvens SAEVETSEG VDESEKKNSG ALEKENQMRM ADLSELLKEG TKEAHDRAEN TQFVKDFLKG NIKKELFKLA TTALYFTYSA LEEEMERNKD HPAFAPLYFP MELHRKEALT KDMEYFFGEN WEEQVQCPKA AQKYVERIHY IGQNEPELLV AHAYTRYMGD LSGGQVLKKV AQRALKLPST GEGTQFYLFE NVDNAQQFKQ LYRARMNALD LNMKTKERIV EEANKAFEYN MQIFNELDQA GSTLARETLE DGFPVHDGKG DMRK
Buffert HMOX2 solution containing 20mM Tris pH 8.0, 1mM DTT and 10% glycerol.
Renhet eller kvalitetsklass Greater than 90% as determined by SDS-PAGE.