missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Homeobox protein MOX-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Homeobox protein MOX-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6354-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062690

  • 38570.00 SEK / 1mg
Estimated Shipment: 03-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001035091.1
Homeobox protein MOX-1 Protein
Research Use Only
MEOX1
Recombinant Protein
E. coli
MDPAASSCMR SLQPPAPVWG CLRNPHSEGN GASGLPHYPP TPFSFHQKPD FLATATAAYP DFSASCLAAT PHSLPQEEHI FTEQHPAFPQ SPNWHFPVSD ARRRPNSGPA GGSKEMGTSS LGLVDTTGGP GDDYGVLGST ANETEKKSSR RRKESSDNQE NRGKPEGSSK ARKERTAFTK EQLRELEAEF AHHNYLTRLR RYEIAVNLDL SERQVKVWFQ NRRMKWKRVK GGQPISPNGQ DPEDGDSTAS PSSE
Greater than 80% as determined by SDS-PAGE.
4222
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers