missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Homer protein homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Homer protein homolog 1, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001264006.1 |
---|---|
Gene ID (Entrez) | 9456 |
Name | Homer protein homolog 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024591
|
enQuireBio™
QP8433-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044591
|
enQuireBio™
QP8433-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16034591
|
enQuireBio™
QP8433-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054591
|
enQuireBio™
QP8433-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064591
|
enQuireBio™
QP8433-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074591
|
enQuireBio™
QP8433-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001264006.1 | |
Homer protein homolog 1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
9456 | |
Research Use Only | |
HOMER1 | |
Recombinant Protein | |
E. coli | |
MGEQPIFSTR AHVFQIDPNT KKNWVPTSKH AVTVSYFYDS TRNVYRIISL DGSKAIINST ITPNMTFTKT SQKFGQWADS RANTVYGLGF SSEHHLSKFA EKFQEFKEAA RLAKEKSQEK MELTSTPSQE SAGGDLQSPL TPESINGTDD ERTPDVTQNS EPRAEPTQNA LPFSHSSAIS KHWEAELATL KGNNAKLTAA LLESTANVKQ WKQQLAAYQE EAERLHKRVT ELECVSSQAN AVHTHKTELN QTIQELEETL KLKEEEIERL KQEIDNAREL QEQRDSLTQK LQEVEIRNKD LEGQLSDLEQ RLEKSQNEQE AFRNNLKTLL EILDGKIFEL TELRDNLAKL LECS | |
Greater than 90% as determined by SDS-PAGE. |