missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human HPR Protein

Product Code. 15989988
Click to view available options
Kvantitet:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15989988

Brand: enQuireBio™ QP123031mg

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

A cDNA sequence encoding the HPR was constructed and used to recombinantly synthesize the protein.

Specifications

Namn HPR Protein
Kvantitet 1 mg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag His
Sekvens MGSSHHHHHH SSGLVPRGSH MGSLYSGNDV TDISDDRFPK PPEIANGYVE HLFRYQCKNY YRLRTEGDGV YTLNDKKQWI NKAVGDKLPE CEAVCGKPKN PANPVQRILG GHLDAKGSFP WQAKMVSHHN LTTGATLINE QWLLTTAKNL FLNHSENATA KDIAPTLTLY VGKKQLVEIE KVVLHPNYHQ VDIGLIKLKQ KVLVNERVMP ICLPSKNYAE VGRVGYVSGW GQSDNFKLTD HLKYVMLPVA DQYDCITHYE GSTCPKWKAP KSPVGVQPIL NEHTFCVGMS KYQEDTCYGD AGSAFAVHDL EEDTWYAAGI LSFDKSCAVA EYGVYVKVTS IQHWVQKTIA EN
Buffert HPR protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.5), 20% glycerol 1mM DTT and 0.15M NaCl.
Renhet eller kvalitetsklass Greater than 85% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.