missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HRV-1A Protein
A DNA sequence encoding the Homo sapiens (Human) HRV-1A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9561-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
HRV-1A Protein | |
Research Use Only | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1 mg | |
Not determined. | |
Recombinant Protein | |
Yeast | |
NPVENYIDEV LNEVLVVPNI KESHHTTSNS APLLDAAETG HTSNVQPEDA IETRYVITSQ TRDEMSIESF LGRSGCVHIS RIKVDYTDYN GQDINFTKWK ITLQEMAQIR RKFELFTYVR FDSEITLVPC IAGRGDDIGH IVMQYMYVPP GAPIPSKRND FSWQSGTNMS IFWQHGQPFP RFSLPFLSIA SAYYMFYDGY DGDNTSSKYG SVVTNDMGTI CSRIVTEKQK HSVVITTHIY HKAKHTKAWC PRPPRAVPYT HSHVTNYMPE TGDVTTAIVR RNTITTA | |
Greater than 90% as determined by SDS-PAGE. |