missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Hyaluronidase PH-20 Protein

A DNA sequence encoding the Homo sapiens (Human) Hyaluronidase PH-20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6719-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055360

  • 26990.00 SEK / 1mg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001167515.1
Hyaluronidase PH-20 Protein
Research Use Only
SPAM1
Recombinant Protein
E. coli
LNFRAPPVIP NVPFLWAWNA PSEFCLGKFD EPLDMSLFSF IGSPRINATG QGVTIFYVDR LGYYPYIDSI TGVTVNGGIP QKISLQDHLD KAKKDITFYM PVDNLGMAVI DWEEWRPTWA RNWKPKDVYK NRSIELVQQQ NVQLSLTEAT EKAKQEFEKA GKDFLVETIK LGKLLRPNHL WGYYLFPDCY NHHYKKPGYN GSCFNVEIKR NDDLSWLWNE STALYPSIYL NTQQSPVAAT LYVRNRVREA IRVSKIPDAK SPLPVFAYTR IVFTDQVLKF LSQDELVYTF GETVALGASG IVIWGTLSIM RSMKSCLLLD NYMETILNPY IINVTLAAKM CSQVLCQEQG VCIRKNWNSS DYLHLNPDNF AIQLEKGGKF TVRGKPTLED LEQFSEKFYC SCYSTLSCKE KADVKDTDAV DVCIADGVCI DAFLKPPMET EEPQIFYNAS PSTLS
Greater than 90% as determined by SDS-PAGE.
6677
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers