missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Hyaluronidase PH-20 Protein
A DNA sequence encoding the Homo sapiens (Human) Hyaluronidase PH-20, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6719-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001167515.1 | |
Hyaluronidase PH-20 Protein | |
Research Use Only | |
SPAM1 | |
Recombinant Protein | |
E. coli | |
LNFRAPPVIP NVPFLWAWNA PSEFCLGKFD EPLDMSLFSF IGSPRINATG QGVTIFYVDR LGYYPYIDSI TGVTVNGGIP QKISLQDHLD KAKKDITFYM PVDNLGMAVI DWEEWRPTWA RNWKPKDVYK NRSIELVQQQ NVQLSLTEAT EKAKQEFEKA GKDFLVETIK LGKLLRPNHL WGYYLFPDCY NHHYKKPGYN GSCFNVEIKR NDDLSWLWNE STALYPSIYL NTQQSPVAAT LYVRNRVREA IRVSKIPDAK SPLPVFAYTR IVFTDQVLKF LSQDELVYTF GETVALGASG IVIWGTLSIM RSMKSCLLLD NYMETILNPY IINVTLAAKM CSQVLCQEQG VCIRKNWNSS DYLHLNPDNF AIQLEKGGKF TVRGKPTLED LEQFSEKFYC SCYSTLSCKE KADVKDTDAV DVCIADGVCI DAFLKPPMET EEPQIFYNAS PSTLS | |
Greater than 90% as determined by SDS-PAGE. |
6677 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |