missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDH3B Protein
A DNA sequence encoding the Homo sapiens (Human) IDH3B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6193-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001317692.1 | |
IDH3B Protein | |
Research Use Only | |
IDH3B | |
Recombinant Protein | |
E. coli | |
ASRSQAEDVR VEGSFPVTML PGDGVGPELM HAVKEVFKAA AVPVEFQEHH LSEVQNMASE EKLEQVLSSM KENKVAIIGK IHTPMEYKGE LASYDMRLRR KLDLFANVVH VKSLPGYMTR HNNLDLVIIR EQTEGEYSSL EHESARGVIE CLKIVTRAKS QRIAKFAFDY ATKKGRGKVT AVHKANIMKL GDGLFLQCCE EVAELYPKIK FETMIIDNCC MQLVQNPYQF DVLVMPNLYG NIIDNLAAGL VGGAGVVPGE SYSAEYAVFE TGARHPFAQA VGRNIANPTA MLLSASNMLR HLNLEYHSSM IADAVKKVIK VGKVRTRDMG GYSTTTDFIK SVIGHLQTKG S | |
Greater than 90% as determined by SDS-PAGE. |
3420 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |