missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IDO2 Protein
A DNA sequence encoding the Homo sapiens (Human) IDO2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6196-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_919270.2 | |
IDO2 Protein | |
Research Use Only | |
IDO2 | |
Recombinant Protein | |
E. coli | |
MEPHRPNVKT AVPLSLESYH ISEEYGFLLP DSLKELPDHY RPWMEIANKL PQLIDAHQLQ AHVDKMPLLS CQFLKGHREQ RLAHLVLSFL TMGYVWQEGE AQPAEVLPRN LALPFVEVSR NLGLPPILVH SDLVLTNWTK KDPDGFLEIG NLETIISFPG GESLHGFILV TALVEKEAVP GIKALVQATN AILQPNQEAL LQALQRLRLS IQDITKTLGQ MHDYVDPDIF YAGIRIFLSG WKDNPAMPAG LMYEGVSQEP LKYSGGSAAQ STVLHAFDEF LGIRHSKESG DFLYRMRDYM PPSHKAFIED IHSAPSLRDY ILSSGQDHLL TAYNQCVQAL AELRSYHITM VTKYLITAAA KAKHGKPNHL PGPPQALKDR GTGGTAVMSF LKSVRDKTLE SILHPRG | |
Greater than 90% as determined by SDS-PAGE. |
169355 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |