Learn More
enQuireBio™ Recombinant Human IDO2 Protein
A DNA sequence encoding the Homo sapiens (Human) IDO2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6196-ec-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16071480
  • 26990.00 SEK / 1mg
Estimated Shipment: 24-06-2024
to see stock.
Specifications
NP_919270.2
IDO2 Protein
Research Use Only
IDO2
Recombinant Protein
E. coli
MEPHRPNVKT AVPLSLESYH ISEEYGFLLP DSLKELPDHY RPWMEIANKL PQLIDAHQLQ AHVDKMPLLS CQFLKGHREQ RLAHLVLSFL TMGYVWQEGE AQPAEVLPRN LALPFVEVSR NLGLPPILVH SDLVLTNWTK KDPDGFLEIG NLETIISFPG GESLHGFILV TALVEKEAVP GIKALVQATN AILQPNQEAL LQALQRLRLS IQDITKTLGQ MHDYVDPDIF YAGIRIFLSG WKDNPAMPAG LMYEGVSQEP LKYSGGSAAQ STVLHAFDEF LGIRHSKESG DFLYRMRDYM PPSHKAFIED IHSAPSLRDY ILSSGQDHLL TAYNQCVQAL AELRSYHITM VTKYLITAAA KAKHGKPNHL PGPPQALKDR GTGGTAVMSF LKSVRDKTLE SILHPRG
Greater than 90% as determined by SDS-PAGE.
169355
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol