missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFNA14 / Interferon alpha-14 Protein

A DNA sequence encoding the Homo sapiens (Human) IFNA14 / Interferon alpha-14, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6200-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061520

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002163.2
IFNA14 / Interferon alpha-14 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3448
Greater than 80% as determined by SDS-PAGE.
Research Use Only
IFNA14
Recombinant Protein
Yeast
CNLSQTHSLN NRRTLMLMAQ MRRISPFSCL KDRHDFEFPQ EEFDGNQFQK AQAISVLHEM MQQTFNLFST KNSSAAWDET LLEKFYIELF QQMNDLEACV IQEVGVEETP LMNEDSILAV KKYFQRITLY LMEKKYSPCA WEVVRAEIMR SLSFSTNLQK RLRRKD
Videos
SDS
Documents