missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IFNG / Interferon Gamma Protein

A DNA sequence encoding the Homo sapiens (Human) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8548-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065211

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000610.2
IFNG / Interferon Gamma Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3458
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFNG
Recombinant Protein
E. coli
QDPYVKEAEN LKKYFNAGHS DVADNGTLFL GILKNWKEES DRKIMQSQIV SFYFKLFKNF KDDQSIQKSV ETIKEDMNVK FFNSNKKKRD DFEKLTNYSV TDLNVQRKAI HELIQVMAEL SPAAKTGKRK RSQMLFRG
Videos
SDS
Documents