missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-1 beta / IL1B Protein

A DNA sequence encoding the Homo sapiens (Human) IL-1 beta / IL1B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6214-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061630

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_000567.1
IL-1 beta / IL1B Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
3553
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL1B
Recombinant Protein
E. coli
APVRSLNCTL RDSQQKSLVM SGPYELKALH LQGQDMEQQV VFSMSFVQGE ESNDKIPVAL GLKEKNLYLS CVLKDDKPTL QLESVDPKNY PKKKMEKRFV FNKIEINNKL EFESAQFPNW YISTSQAENM PVFLGGTKGG QDITDFTMQF VSS
Videos
SDS
Documents