missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 23 Protein
A cDNA sequence encoding the IL 23 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10704-10ug
Additional Details : Weight : 0.01000kg
Specifications
51561 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
The activity, as determined by the dose dependent secretion of IL-17 in response to IL-23 using murine splenocytes, the ED50 is 1.9ng/ml. | |
Human | |
Untagged | |
IL-23 was lyophilized from a 0.1microm filtered solution (1.05 mg/ml) containing 1xPBS. |
IL 23 Protein | |
Research Use Only | |
IL23A | |
Recombinant Protein | |
Human (HEK293) | |
MLGSRAVMLL LLLPWTAQGR AVPGGSSPAW TQCQQLSQKL CTLAWSAHPL VGHMDLREEG DEETTNDVPH IQCGDGCDPQ GLRDNSQFCL QRIHQGLIFY EKLLGSDIFT GEPSLLPDSP VGQLHASLLG LSQLLQPEGH HWETQQIPSL SPSQPWQRLL LRFKILRSLQ AFVAVAARVF AHGAATLSP | |
Greater than 95.0% as determined by analysis by SDS-PAGE. |