missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-29 / Interleukin-29 Protein
A DNA sequence encoding the Homo sapiens (Human) IL-29 / Interleukin-29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7814-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_742152.1 | |
IL-29 / Interleukin-29 Protein | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
282618 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IFNL1 | |
Recombinant Protein | |
E. coli | |
PVPTSKPTTT GKGCHIGRFK SLSPQELASF KKARDALEES LKLKNWSCSS PVFPGNWDLR LLQVRERPVA LEAELALTLK VLEAAAGPAL EDVLDQPLHT LHHILSQLQA CIQPQPTAGP RPRGRLHHWL HRLQEAPKKE SAGCLEASVT FNLFRLLTRD LKYVADGNLC LRTSTHPEST |