missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL12RB2 / IL12R-beta 2 Protein
A DNA sequence encoding the Homo sapiens (Human) IL12RB2 / IL12R-beta 2, was expressed in the hosts and tags indicated.
2005.00 SEK - 22130.00 SEK
Specifications
Accession Number | NP_001245143.1 |
---|---|
Gene ID (Entrez) | 3595 |
Name | IL12RB2 / IL12R-beta 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068901
|
enQuireBio™
QP9881-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008911
|
enQuireBio™
QP9881-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078901
|
enQuireBio™
QP9881-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098901
|
enQuireBio™
QP9881-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018911
|
enQuireBio™
QP9881-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088901
|
enQuireBio™
QP9881-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001245143.1 | |
IL12RB2 / IL12R-beta 2 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3595 | |
Research Use Only | |
IL12RB2 | |
Recombinant Protein | |
Yeast | |
KIDACKRGDV TVKPSHVILL GSTVNITCSL KPRQGCFHYS RRNKLILYKF DRRINFHHGH SLNSQVTGLP LGTTLFVCKL ACINSDEIQI CGAEIFVGVA PEQPQNLSCI QKGEQGTVAC TWERGRDTHL YTEYTLQLSG PKNLTWQKQC KDIYCDYLDF GINLTPESPE SNFTAKVTAV NSLGSSSSLP STFTFLDIVR PLPPWDIRIK FQKASVSRCT LYWRDEGLVL LNRLRYRPSN SRLWNMVNVT KAKGRHDLLD LKPFTEYEFQ ISSKLHLYKG SWSDWSESLR AQTPEEEPTG MLDVWYMKRH IDYSRQQISL FWKNLSVSEA RGKILHYQVT LQELTGGKAM TQNITGHTSW TTVIPRTGNW AVAVSAANSK GSSLPTRINI MNLCEAGLLA PRQVSANSEG MDNILVTWQP PRKDPSAVQE YVVEWRELHP GGDTQVPLNW LRSRPYNVSA LISENIKSYI CYEIRVYALS GDQGGCSSIL GNSKHKAPLS GPHINAITEE KGSILISWNS IPVQEQMGCL LHYRIYWKER DSNSQPQLCE IPYRVSQNSH PINSLQPRVT YVLWMTALTA AGESSHGNER EFCLQGKAN | |
Greater than 90% as determined by SDS-PAGE. |