missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL1F5 / IL36RN Protein
A DNA sequence encoding the Homo sapiens (Human) IL1F5 / IL36RN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8080-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036407.1 | |
IL1F5 / IL36RN Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
26525 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
IL36RN | |
Recombinant Protein | |
E. coli | |
MVLSGALCFR MKDSALKVLY LHNNQLLAGG LHAGKVIKGE EISVVPNRWL DASLSPVILG VQGGSQCLSC GVGQEPTLTL EPVNIMELYL GAKESKSFTF YRRDMGLTSS FESAAYPGWF LCTVPEADQP VRLTQLPENG GWNAPITDFY FQQCD |