missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL1F6 / IL36 Protein

A DNA sequence encoding the Homo sapiens (Human) IL1F6 / IL36, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8208-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063281

  • 26990.00 SEK / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_055255.1
IL1F6 / IL36 Protein
Research Use Only
Il36a
Recombinant Protein
E. coli
MEKALKIDTP QQGSIQDINH RVWVLQDQTL IAVPRKDRMS PVTIALISCR HVETLEKDRG NPIYLGLNGL NLCLMCAKVG DQPTLQLKEK DIMDLYNQPE PVKSFLFYHS QSGRNSTFES VAFPGWFIAV SSEGGCPLIL TQELGKANTT DFGLTMLF
Greater than 80% as determined by SDS-PAGE.
27179
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers