missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Immunity-related GTPase family M Protein
A DNA sequence encoding the Homo sapiens (Human) Immunity-related GTPase family M protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6236-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139277.1 | |
Immunity-related GTPase family M Protein | |
Research Use Only | |
IRGM | |
Recombinant Protein | |
E. coli | |
MEAMNVEKAS ADGNLPEVIS NIKETLKIVS RTPVNITMAG DSGNGMSTFI SALRNTGHEG KASPPTELVK ATQRCASYFS SHFSNVVLWD LPGTGSATTT LENYLMEMQF NRYDFIMVAS AQFSMNHVML AKTAEDMGKK FYIVWTKLDM DLSTGALPEV QLLQIRENVL ENLQKERVCE Y | |
Greater than 80% as determined by SDS-PAGE. |
345611 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |