missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Inactive tyrosine-protein kinase 7 Protein

A DNA sequence encoding the Homo sapiens (Human) Inactive tyrosine-protein kinase 7, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9763-ye-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058681

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001257327.1
Inactive tyrosine-protein kinase 7 Protein
10 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
5754
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PTK7
Recombinant Protein
Yeast
AIVFIKQPSS QDALQGRRAL LRCEVEAPGP VHVYWLLDGA PVQDTERRFA QGSSLSFAAV DRLQDSGTFQ CVARDDVTGE EARSANASFN IKWIEAGPVV LKHPASEAEI QPQTQVTLRC HIDGHPRPTY QWFRDGTPLS DGQSNHTVSS KERNLTLRPA GPEHSGLYSC CAHSAFGQAC SSQNFTLSIA DESFARVVLA PQDVVVARYE EAMFHCQFSA QPPPSLQWLF EDETPITNRS RPPHLRRATV FANGSLLLTQ VRPRNAGIYR CIGQGQRGPP IILEATLHLA EIEDMPLFEP RVFTAGSEER VTCLPPKGLP EPSVWWEHAG VRLPTHGRVY QKGHELVLAN IAESDAGVYT CHAANLAGQR RQDVNITVAT VPSWLKKPQD SQLEEGKPGY LDCLTQATPK PTVVWYRNQM LISEDSRFEV FKNGTLRINS VEVYDGTWYR CMSSTPAGSI EAQARVQVLE KLKFTPPPQP QQCMEFDKEA TVPCSATGRE KPTIKWERAD GSSLPEWVTD NAGTLHFARV TRDDAGNYTC IASNGPQGQI RAHVQLTVAV FITFKVEPER TTVYQGHTAL LQCEAQGDPK PLIQWKGKDR ILDPTKLGPR MHIFQNGSLV IHDVAPEDSG RYTCIAGNSC NIKHTEAPLY VVDKPVPEES EGPGSPPPYK MIQT
Videos
SDS
Documents