missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inactive tyrosine-protein kinase 7 Protein
A DNA sequence encoding the Homo sapiens (Human) Inactive tyrosine-protein kinase 7, was expressed in the hosts and tags indicated.
2005.00 SEK - 22130.00 SEK
Specifications
Accession Number | NP_001257327.1 |
---|---|
Gene ID (Entrez) | 5754 |
Name | Inactive tyrosine-protein kinase 7 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048681
|
enQuireBio™
QP9763-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088681
|
enQuireBio™
QP9763-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058681
|
enQuireBio™
QP9763-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078681
|
enQuireBio™
QP9763-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098681
|
enQuireBio™
QP9763-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068681
|
enQuireBio™
QP9763-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257327.1 | |
Inactive tyrosine-protein kinase 7 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5754 | |
Research Use Only | |
PTK7 | |
Recombinant Protein | |
Yeast | |
AIVFIKQPSS QDALQGRRAL LRCEVEAPGP VHVYWLLDGA PVQDTERRFA QGSSLSFAAV DRLQDSGTFQ CVARDDVTGE EARSANASFN IKWIEAGPVV LKHPASEAEI QPQTQVTLRC HIDGHPRPTY QWFRDGTPLS DGQSNHTVSS KERNLTLRPA GPEHSGLYSC CAHSAFGQAC SSQNFTLSIA DESFARVVLA PQDVVVARYE EAMFHCQFSA QPPPSLQWLF EDETPITNRS RPPHLRRATV FANGSLLLTQ VRPRNAGIYR CIGQGQRGPP IILEATLHLA EIEDMPLFEP RVFTAGSEER VTCLPPKGLP EPSVWWEHAG VRLPTHGRVY QKGHELVLAN IAESDAGVYT CHAANLAGQR RQDVNITVAT VPSWLKKPQD SQLEEGKPGY LDCLTQATPK PTVVWYRNQM LISEDSRFEV FKNGTLRINS VEVYDGTWYR CMSSTPAGSI EAQARVQVLE KLKFTPPPQP QQCMEFDKEA TVPCSATGRE KPTIKWERAD GSSLPEWVTD NAGTLHFARV TRDDAGNYTC IASNGPQGQI RAHVQLTVAV FITFKVEPER TTVYQGHTAL LQCEAQGDPK PLIQWKGKDR ILDPTKLGPR MHIFQNGSLV IHDVAPEDSG RYTCIAGNSC NIKHTEAPLY VVDKPVPEES EGPGSPPPYK MIQT | |
Greater than 90% as determined by SDS-PAGE. |