missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inosine-5'-monophosphate dehydrogenase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Inosine-5'-monophosphate dehydrogenase 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8353-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000875.2 | |
Inosine-5'-monophosphate dehydrogenase 2 Protein | |
Research Use Only | |
IMPD2 | |
Recombinant Protein | |
E. coli | |
LISGGTSYVP DDGLTAQQLF NCGDGLTYND FLILPGYIDF TADQVDLTSA LTKKITLKTP LVSSPMDTVT EAGMAIAMAL TGGIGFIHHN CTPEFQANEV RKVKKYEQGF ITDPVVLSPK DRVRDVFEAK ARHGFCGIPI TDTGRMGSRL VGIISSRDID FLKEEEHDCF LEEIMTKRED LVVAPAGITL KEANEILQRS KKGKLPIVNE DDELVAIIAR TDLKKNRDYP LASKDAKKQL LCGAAIGTHE DDKYRLDLLA QAGVDVVVLD SSQGNSIFQI NMIKYIKDKY PNLQVIGGNV VTAAQAKNLI DAGVDALRVG MGSGSICITQ EVLACGRPQA TAVYKVSEYA RRFGVPVIAD GGIQNVGHIA KALALGASTV MMGSLLAATT EAPGEYFFSD GIRLKKYRGM GSLDAMDKHL SSQNRYFSEA DKIKVAQGVS GAVQDKGSIH KFVPYLIAGI QHSCQDIGAK SLTQVRAMMY SGELKFEKRT SSAQVEGGVH SLHSYEKRLF | |
Greater than 90% as determined by SDS-PAGE. |
3615 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |