missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inositol-trisphosphate 3-kinase B Protein
A DNA sequence encoding the Homo sapiens (Human) Inositol-trisphosphate 3-kinase B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6243-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002212.3 | |
Inositol-trisphosphate 3-kinase B Protein | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3707 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ITPKB | |
Recombinant Protein | |
E. coli | |
RVEGGSPTLG LLGGSPSAQP GTGNVEAGIP SGRMLEPLPC WDAAKDLKEP QCPPGDRVGV QPGNSRVWQG TMEKAGLAWT RGTGVQSEGT WESQRQDSDA LPSPELLPQD PDKPFLRKAC SPSNIPAVII TDMGTQEDGA LEETQGSPRG NLPLRKLSSS SASSTGFSSS YEDSEEDISS DPERTLDPNS AFLHTLDQQK PRVSKSWRKI KNMVHWSPFV MSFKKKYPWI QLAGHAGSFK AAANGRILKK HCESEQRCLD RLMVDVLRPF VPAYHGDVVK DGERYNQMDD LLADFDSPCV MDCKMGIRTY LEEELTKARK KPSLRKDMYQ KMIEVDPEAP TEEEKAQRAV TKPRYMQWRE TISSTATLGF RIEGIKKEDG TVNRDFKKTK TREQVTEAFR EFTKGNHNIL IAYRDRLKAI RTTLEVSPFF KCHEVIGSSL LFIHDKKEQA KVWMIDFGKT TPLPEGQTLQ HDVPWQEGNR EDGYLSGLNN LVDILTEMSQ DAPLA |