missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin alpha-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6238-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002194.2 | |
Integrin alpha-2 Protein | |
Research Use Only | |
ITGA2 | |
Recombinant Protein | |
Yeast | |
WDAVKNFLEK FVQGLDIGPT KTQVGLIQYA NNPRVVFNLN TYKTKEEMIV ATSQTSQYGG DLTNTFGAIQ YARKYAYSAA SGGRRSATKV MVVVTDGESH DGSMLKAVID QCNHDNILRF GIAVLGYLNR NALDTKNLIK EIKAIASIPT ERYFFNVSDE AALLEKAGTL GEQIFSIE | |
Greater than 90% as determined by SDS-PAGE. |
3673 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |