missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Integrin alpha-IIb Protein
A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-IIb, was expressed in the hosts and tags indicated.
2005.00 SEK - 22130.00 SEK
Specifications
Accession Number | NP_000410.2 |
---|---|
Gene ID (Entrez) | 3674 |
Name | Integrin alpha-IIb Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051840
|
enQuireBio™
QP6239-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16091840
|
enQuireBio™
QP6239-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061840
|
enQuireBio™
QP6239-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081840
|
enQuireBio™
QP6239-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001850
|
enQuireBio™
QP6239-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16071840
|
enQuireBio™
QP6239-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000410.2 | |
Integrin alpha-IIb Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3674 | |
Research Use Only | |
ITGA2B | |
Recombinant Protein | |
Yeast | |
CVPQLQLTAS VTGSPLLVGA DNVLELQMDA ANEGEGAYEA ELAVHLPQGA HYMRALSNVE GFERLICNQK KENETRVVLC ELGNPMKKNA QIGIAMLVSV GNLEEAGESV SFQLQIRSKN SQNPNSKIVL LDVPVRAEAQ VELRGNSFPA SLVVAAEEGE REQNSLDSWG PKVEHTYELH NNGPGTVNGL HLSIHLPGQS QPSDLLYILD IQPQGGLQCF PQPPVNPLKV DWGLPIPSPS PIHPAHHKR | |
Greater than 90% as determined by SDS-PAGE. |