missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Inter-alpha-trypsin inhibitor heavy chain H5 Protein
A DNA sequence encoding the Homo sapiens (Human) Inter-alpha-trypsin inhibitor heavy chain H5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7779-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001001851.1 | |
Inter-alpha-trypsin inhibitor heavy chain H5 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
80760 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ITIH5 | |
Recombinant Protein | |
E. coli | |
VPRQVRLLQR LKTKPLMTEF SVKSTIISRY AFTTVSCRML NRASEDQDIE FQMQIPAAAF ITNFTMLIGD KVYQGEITER EKKSGDRVKE KRNKTTEENG EKGTEIFRAS AVIPSKDKAA FFLSYEE |