missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interferon alpha-21 Protein

A DNA sequence encoding the Homo sapiens (Human) Interferon alpha-21, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6201-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061530

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_002166.2
Interferon alpha-21 Protein
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
3452
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFNA21
Recombinant Protein
Yeast
CDLPQTHSLG NRRALILLAQ MGRISPFSCL KDRHDFGFPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST KDSSATWEQS LLEKFSTELN QQLNDLEACV IQEVGVEETP LMNVDSILAV KKYFQRITLY LTEKKYSPCA WEVVRAEIMR SFSLSKIFQE RLRRKE
Videos
SDS
Documents