missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Interferon alpha 2b Protein
A DNA sequence encoding the Homo sapiens (Human) Interferon alpha 2b, was expressed in the hosts and tags indicated.
2005.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_000596.2 |
---|---|
Gene ID (Entrez) | 3440 |
Name | Interferon alpha 2b Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008350
|
enQuireBio™
QP7345-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16048350
|
enQuireBio™
QP7345-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018350
|
enQuireBio™
QP7345-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038350
|
enQuireBio™
QP7345-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058350
|
enQuireBio™
QP7345-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16028350
|
enQuireBio™
QP7345-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000596.2 | |
Interferon alpha 2b Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3440 | |
Research Use Only | |
IFNA2 | |
Recombinant Protein | |
Yeast | |
CDLPQTHSLG SRRTLMLLAQ MRKISLFSCL KDRHDFGFPQ EEFGNQFQKA ETIPVLHEMI QQIFNLFSTK DSSAAWDETL LDKFYTELYQ QLNDLEACVI QGVGVTETPL MKEDSILAVR KYFQRITLYL KEKKYSPCAW EVVRAEIMRS FSLSTNLQES LRSKE | |
Greater than 90% as determined by SDS-PAGE. |