missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Intestine-specific homeobox Protein
A DNA sequence encoding the Homo sapiens (Human) Intestine-specific homeobox, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6237-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001290437.1 | |
Intestine-specific homeobox Protein | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
91464 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ISX | |
Recombinant Protein | |
E. coli | |
MCAEVGPALC RGMERNSLGC CEAPKKLSLS FSIEAILKRP ARRSDMDRPE GPGEEGPGEA AASGSGLEKP PKDQPQEGRK SKRRVRTTFT TEQLHELEKI FHFTHYPDVH IRSQLAARIN LPEARVQIWF QNQRAKWRKQ EKIGNLGAPQ QLSEASVALP TNLDVAGPTW TSTALRRLAP PTSCCPSAQD QLASAWFPAW ITLLPAHPWE TQPVPGLPIH QTCIPVLCIL PPPHPKWGSI CATST |