missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Intraflagellar transport protein 22 homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 22 homolog, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8067-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001124292.1 | |
Intraflagellar transport protein 22 homolog Protein | |
Research Use Only | |
RABL5 | |
Recombinant Protein | |
E. coli | |
MLKAKILFVG PCESGKTVLA NFLTESSDIT EYSPTQGVRI LEFENPHVTS NNKGTGCEFE LWDCGGDAKF ESCWPALMKD AHGVVIVFNA DIPSHRKEME MWYSCFVQQP SLQDTQCMLI AHHKPGSGDD KGSLSLSPPL NKLKLVHSNL EDDPEEIRME FIKYLKSIIN SMSESRDREE MSIMT | |
Greater than 90% as determined by SDS-PAGE. |
64792 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |