missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Intraflagellar transport protein 27 homolog Protein

A DNA sequence encoding the Homo sapiens (Human) Intraflagellar transport protein 27 homolog, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8131-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062821

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001171172.1
Intraflagellar transport protein 27 homolog Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
11020
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IFT27
Recombinant Protein
E. coli
MVKLAAKCIL AGDPAVGKTA LAQIFRSDGA HFQKSYTLTT GMDLVVKTVP VPDTGDSVEL FIFDSAGKEL FSEMLDKLWE SPNVLCLVYD VTNEESFNNC SKWLEKARSQ APGISLPGVL VGNKTDLAGR RAVDSAEARA WALGQGLECF ETSVKEMENF EAPFHCLAKQ FHQLYREKVE VFRALA
Videos
SDS
Documents