missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Islet cell autoantigen 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Islet cell autoantigen 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6188-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001129492.1 | |
Islet cell autoantigen 1 Protein | |
Research Use Only | |
ICA1 | |
Recombinant Protein | |
E. coli | |
MSGHKCSYPW DLQDRYAQDK SVVNKMQQKY WETKQAFIKA TGKKEDEHVV ASDADLDAKL ELFHSIQRTC LDLSKAIVLY QKRICFLSQE ENELGKFLRS QGFQDKTRAG KMMQATGKAL CFSSQQRLAL RNPLCRFHQE VETFRHRAIS DTWLTVNRME QCRTEYRGAL LWMKDVSQEL DPDLYKQMEK FRKVQTQVRL AKKNFDKLKM DVCQKVDLLG ASRCNLLSHM LATYQTTLLH FWEKTSHTMA AIHESFKGYQ PYEFTTLK | |
Greater than 90% as determined by SDS-PAGE. |
3382 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |