missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCND1 Protein
A DNA sequence encoding the Homo sapiens (Human) KCND1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6253-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004970.3 | |
KCND1 Protein | |
500 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3750 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
KCND1 | |
Recombinant Protein | |
Yeast | |
NFSRIYHQNQ RADKRRAQQK VRLARIRLAK SGTTNAFLQY KQNGGLEDSG SGEEQALCVR NRSAFEQQHH HLLHCLEKTT CHEFTDELTF SEALGAVSPG GRTSRSTSVS SQPVGPGSLL SSCCPRRAKR RAIRLANSTA SVSRGSMQEL DMLAGLRRSH APQSRSSLNA KPHDSLDLNC DSRDFVAAII SIPTPPANTP DESQPSSPGG GGRAGSTLRN SSLGTPCLFP ETVKISSL |