missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ketohexokinase Protein
A DNA sequence encoding the Homo sapiens (Human) Ketohexokinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6260-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000212.1 | |
Ketohexokinase Protein | |
Research Use Only | |
KHK | |
Recombinant Protein | |
E. coli | |
MEEKQILCVG LVVLDVISLV DKYPKEDSEI RCLSQRWQRG GNASNSCTVL SLLGAPCAFM GSMAPGHVAD FVLDDLRRYS VDLRYTVFQT TGSVPIATVI INEASGSRTI LYYDRSLPDV SATDFEKVDL TQFKWIHIEG RNASEQVKML QRIDAHNTRQ PPEQKIRVSV EVEKPREELF QLFGYGDVVF VSKDVAKHLG FQSAEEALRG LYGRVRKGAV LVCAWAEEGA DALGPDGKLL HSDAFPPPRV VDTLGAGDTF NASVIFSLSQ GRSVQEALRF GCQVAGKKCG LQGFDGIV | |
Greater than 80% as determined by SDS-PAGE. |
3795 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |