missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kidney mitochondrial carrier protein 1
A DNA sequence encoding the Homo sapiens (Human) Kidney mitochondrial carrier protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7723-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001010875.1 | |
Kidney mitochondrial carrier protein 1 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
253512 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SLC25A30 | |
Recombinant Protein | |
E. coli | |
MSALNWKPFV YGGLASITAE CGTFPIDLTK TRLQIQGQTN DAKFKEIRYR GMLHALVRIG REEGLKALYS GIAPAMLRQA SYGTIKIGTY QSLKRLFIER PEDETLPINV ICGILSGVIS STIANPTDVL KIRMQAQSNT IQGGMIGNFM NIYQQEGTRG LWKGVSLTAQ RAAIVVGVEL PVYDITKKHL ILSGLMGDTV YTHFLSSFTC GLAGALASNP VDVVRTRMMN QRVLRDGRCS GYTGTLDCLL QTWKNEGFFA LYKGFWPNWL RLGPWNIIFF VTYEQLKKLD L |