missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLHDC8B Protein
A cDNA sequence encoding the KLHDC8B was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | KLHDC8B Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15970619
|
enQuireBio™
QP12501-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15960619
|
enQuireBio™
QP12501-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950619
|
enQuireBio™
QP12501-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
KLHDC8B Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KLHDC8B protein solution (0.5 mg/ml) containing Phosphate buffer saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSAGGGR AFAWQVFPPM PTCRVYGTVA HQDGHLLVLG GCGRAGLPLD TAETLDMASH TWLALAPLPT ARAGAAAVVL GKQVLVVGGV DEVQSPVAAV EAFLMDEGRW ERRATLPQAA MGVATVERDG MVYALGGMGP DTAPQAQVRV YEPRRDCWLS LPSMPTPCYG ASTFLHGNKI YVLGGRQGKL PVTAFEAFDL EARTWTRHPS LPSRRAFAGC AMAEGSVFSL GGLQQPGPHN FYSRPHFVNT VEMFDLEHGS WTKLPRSLRM RDKRADFVVG SLGGHIVAIG GLGNQPCPLG SVESFSLARR RWEALPAMPT ARCSCSSLQA GPRLFVIGGV AQGPSQAVEA LCLRDGV | |
Greater than 85.0% as determined by SDS-PAGE. |