missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLK7 / PRSS6 Protein

A DNA sequence encoding the Homo sapiens (Human) KLK7 / PRSS6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6267-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062060

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001193982.1
KLK7 / PRSS6 Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
5650
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KLK7
Recombinant Protein
Yeast
DKIIDGAPCA RGSHPWQVAL LSGNQLHCGG VLVNERWVLT AAHCKMNEYT VHLGSDTLGD RRAQRIKASK SFRHPGYSTQ THVNDLMLVK LNSQARLSSM VKKVRLPSRC EPPGTTCTVS GWGTTTSPDV TFPSDLMCVD VKLISPQDCT KVYKDLLENS MLCAGIPDSK KNACNGDSGG PLVCRGTLQG LVSWGTFPCG QPNDPGVYTQ VCKFTKWIND TMKKHR
Videos
SDS
Documents