missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KLRG2 Protein
A DNA sequence encoding the Homo sapiens (Human) KLRG2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9236-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_940910.1 | |
KLRG2 Protein | |
Research Use Only | |
KLRG2 | |
Recombinant Protein | |
Yeast | |
MEESWEAAPG GQAGAELPME PVGSLVPTLE QPQVPAKVRQ PEGPESSPSP AGAVEKAAGA GLEPSSKKKP PSPRPGSPRV PPLSLGYGVC PEPPSPGPAL VKLPRNGEAP GAEPAPSAWA PMELQVDVRV KPVGAAGGSS TPSPRPSTRF LKVPVPESPA FSRHADPAHQ LLLRAPSQGG TWGRRSPLAA ARTESGCDAE GRASPAEGSA GSPGSPTCCR CKELGLEKED AALLPRAGLD GDEKLPRAVT LTGLPMYVKS LYWALAFMAV LLAVSGVVIV VLASRAGARC QQCPPGWVLS EEHCYYFSAE AQAWEASQAF CSAYHATLPL LSHTQDFLGR YPVSRHSWVG AWRGPQGWHW IDEAPLPPQL LPEDGEDNLD INCGALEEGT LVAANCSTPR PWVCAKGTQ | |
Greater than 90% as determined by SDS-PAGE. |
346689 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |