missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human KLRG2 Protein

A DNA sequence encoding the Homo sapiens (Human) KLRG2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9236-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16037891

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_940910.1
KLRG2 Protein
Research Use Only
KLRG2
Recombinant Protein
Yeast
MEESWEAAPG GQAGAELPME PVGSLVPTLE QPQVPAKVRQ PEGPESSPSP AGAVEKAAGA GLEPSSKKKP PSPRPGSPRV PPLSLGYGVC PEPPSPGPAL VKLPRNGEAP GAEPAPSAWA PMELQVDVRV KPVGAAGGSS TPSPRPSTRF LKVPVPESPA FSRHADPAHQ LLLRAPSQGG TWGRRSPLAA ARTESGCDAE GRASPAEGSA GSPGSPTCCR CKELGLEKED AALLPRAGLD GDEKLPRAVT LTGLPMYVKS LYWALAFMAV LLAVSGVVIV VLASRAGARC QQCPPGWVLS EEHCYYFSAE AQAWEASQAF CSAYHATLPL LSHTQDFLGR YPVSRHSWVG AWRGPQGWHW IDEAPLPPQL LPEDGEDNLD INCGALEEGT LVAANCSTPR PWVCAKGTQ
Greater than 90% as determined by SDS-PAGE.
346689
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers