missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Kynurenine--oxoglutarate transaminase 3 Protein
A DNA sequence encoding the Homo sapiens (Human) Kynurenine--oxoglutarate transaminase 3, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001008661.1 |
---|---|
Gene ID (Entrez) | 56267 |
Name | Kynurenine--oxoglutarate transaminase 3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16040521
|
enQuireBio™
QP7739-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060521
|
enQuireBio™
QP7739-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16050521
|
enQuireBio™
QP7739-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16070521
|
enQuireBio™
QP7739-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080521
|
enQuireBio™
QP7739-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090521
|
enQuireBio™
QP7739-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001008661.1 | |
Kynurenine--oxoglutarate transaminase 3 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56267 | |
Research Use Only | |
CCBL | |
Recombinant Protein | |
E. coli | |
MFLAQRSLCS LSGRAKFLKT ISSSKILGFS TSAKMSLKFT NAKRIEGLDS NVWIEFTKLA ADPSVVNLGQ GFPDISPPTY VKEELSKIAA IDSLNQYTRG FGHPSLVKAL SYLYEKLYQK QIDSNKEILV TVGAYGSLFN TIQALIDEGD EVILIVPFYD CYEPMVRMAG ATPVFIPLRS KPVYGKRWSS SDWTLDPQEL ESKFNSKTKA IILNTPHNPL GKVYNREELQ VIADLCIKYD TLCISDEVYE WLVYSGNKHL KIATFPGMWE RTITIGSAGK TFSVTGWKLG WSIGPNHLIK HLQTVQQNTI YTCATPLQEA LAQAFWIDIK RMDDPECYFN SLPKELEVKR DRMVRLLESV GLKPIVPDGG YFIIADVSLL DPDLSDMKNN EPYDYKFVKW MTKHKKLSAI PVSAFCNSET KSQFEKFVRF CFIKKDSTLD AAEEIIKAWS VQKS | |
Greater than 90% as determined by SDS-PAGE. |