Läs mer
R&D Systems™ Recombinant Human LBP Protein
Formulering:
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA.
Beskrivning
The Recombinant Human LBP Protein is derived from NS0. The Recombinant Human LBP Protein has been validated for the following applications: Bioactivity.
Specifikationer
Specifikationer
| Formulering | Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA. |
| Gen-ID (Entrez) | 3929 |
| Molekylvikt (g/mol) | 51 kDa |
| Namn | LBP |
| Kvantitet | 25 μg |
| Källa | Mouse myeloma cell line,NS0-derived human LBP protein Ala26-Val481 |
| Förvaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Konjugera | Unconjugated |
| Rekombinant | Recombinant |
| Renhet eller kvalitetsklass | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |