missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LCAT Protein
A DNA sequence encoding the Homo sapiens (Human) LCAT, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_000220.1 |
---|---|
Gene ID (Entrez) | 3931 |
Name | LCAT Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046081
|
enQuireBio™
QP8712-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066081
|
enQuireBio™
QP8712-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16056081
|
enQuireBio™
QP8712-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076081
|
enQuireBio™
QP8712-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086081
|
enQuireBio™
QP8712-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096081
|
enQuireBio™
QP8712-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000220.1 | |
LCAT Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3931 | |
Research Use Only | |
LCAT | |
Recombinant Protein | |
E. coli | |
FWLLNVLFPP HTTPKAELSN HTRPVILVPG CLGNQLEAKL DKPDVVNWMC YRKTEDFFTI WLDLNMFLPL GVDCWIDNTR VVYNRSSGLV SNAPGVQIRV PGFGKTYSVE YLDSSKLAGY LHTLVQNLVN NGYVRDETVR AAPYDWRLEP GQQEEYYRKL AGLVEEMHAA YGKPVFLIGH SLGCLHLLYF LLRQPQAWKD RFIDGFISLG APWGGSIKPM LVLASGDNQG IPIMSSIKLK EEQRITTTSP WMFPSRMAWP EDHVFISTPS FNYTGRDFQR FFADLHFEEG WYMWLQSRDL LAGLPAPGVE VYCLYGVGLP TPRTYIYDHG FPYTDPVGVL YEDGDDTVAT RSTELCGLWQ GRQPQPVHLL PLHGIQHLNM VFSNLTLEHI NAILLGAYRQ GPPASPTAS | |
Greater than 90% as determined by SDS-PAGE. |