missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Legumain / LGMN Protein
A DNA sequence encoding the Homo sapiens (Human) Legumain/ LGMN, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6301-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001008530.1 | |
Legumain / LGMN Protein | |
Research Use Only | |
Lgmn | |
Recombinant Protein | |
Yeast | |
VPIDDPEDGG KHWVVIVAGS NGWYNYRHQA DACHAYQIIH RNGIPDEQIV VMMYDDIAYS EDNPTPGIVI NRPNGTDVYQ GVPKDYTGED VTPQNFLAVL RGDAEAVKGI GSGKVLKSGP QDHVFIYFTD HGSTGILVFP NEDLHVKDLN ETIHYMYKHK MYRKMVFYIE ACESGSMMNH LPDNINVYAT TAANPRESSY ACYYDEKRST YLGDWYSVNW MEDSDVEDLT KETLHKQYHL VKSHTNTSHV MQYGNKTIST MKVMQFQGMK RKASSPVPLP PVTHLDLTPS PDVPLTIMKR KLMNTN | |
Greater than 90% as determined by SDS-PAGE. |
5641 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |