missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LFNG Protein

A DNA sequence encoding the Homo sapiens (Human) LFNG, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7905-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061511

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001035257.1
LFNG Protein
50 μg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
3955
Greater than 80% as determined by SDS-PAGE.
Research Use Only
LFNG
Recombinant Protein
E. coli
MTPGRCCLAA DIQVETFIFT DGEDEALARH TGNVVITNCS AAHSRQALSC KMAVEYDRFI ESGRKWFCHV DDDNYVNLRA LLRLLASYPH TRDVYVGKPS LDRPIQAMER VSENKVRPVH FWFATGGAGF CISRGLALKM SPWASGGHFM NTAERIRLPD DCTIGYIVEA LLGVPLIRSG LFHSHLENLQ QVPTSELHEQ VTLSYGMFEN KRNAVHVKGP FSVEADPSRF RSIHCHLYPD TPWCPRTAIF
Videos
SDS
Documents