missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LFNG Protein
A DNA sequence encoding the Homo sapiens (Human) LFNG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7905-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035257.1 | |
LFNG Protein | |
Research Use Only | |
LFNG | |
Recombinant Protein | |
E. coli | |
MTPGRCCLAA DIQVETFIFT DGEDEALARH TGNVVITNCS AAHSRQALSC KMAVEYDRFI ESGRKWFCHV DDDNYVNLRA LLRLLASYPH TRDVYVGKPS LDRPIQAMER VSENKVRPVH FWFATGGAGF CISRGLALKM SPWASGGHFM NTAERIRLPD DCTIGYIVEA LLGVPLIRSG LFHSHLENLQ QVPTSELHEQ VTLSYGMFEN KRNAVHVKGP FSVEADPSRF RSIHCHLYPD TPWCPRTAIF | |
Greater than 80% as determined by SDS-PAGE. |
3955 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |