missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LRBA Protein
A DNA sequence encoding the Homo sapiens (Human) LRBA, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6318-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001186211.2 | |
LRBA Protein | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
987 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LRBA | |
Recombinant Protein | |
E. coli | |
PQPHRHVLEI SRQHEQPGQG IAPDAVNGQR RDSRSTVFRI PEFNWSQMHQ RLLTDLLFSI ETDIQMWRSH STKTVMDFVN SSDNVIFVHN TIHLISQVMD NMVMACGGIL PLLSAATSAT HELENIEPTQ GLSIEASVTF LQRLISLVDV LIFASSLGFT EIEAEKSMSS GGILRQCLRL VCAVAVRNCL ECQQHSQLKT RGDKALKPMH SLIPLGKSAA KSPVDIVTGG ISPV |